Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130726.2 | NP_001124198.1 | 4 | 381 | ||
NM_173463.4 | NP_775734.2 | 5 | 482 | |||
XM_005248210.3 | XP_005248267.1 | 4 | 410 | |||
XM_011513906.2 | XP_011512208.1 | 6 | 492 | |||
XM_011513907.2 | XP_011512209.1 | 5 | 365 | |||
XM_011513908.1 | XP_011512210.1 | 4 | 287 | |||
XM_017008827.1 | XP_016864316.1 | 6 | 492 | |||
XM_017008828.1 | XP_016864317.1 | 5 | 416 | |||
GenBank mRNA | AK125353.1 | - | 4 | 290 | ||
AK296454.1 | - | 1 | 39 | |||
AK302905.1 | - | 5 | 426 | |||
AL834257.2 | - | 4 | 355 | |||
BC057761.1 | - | 4 | 357 | |||
BC067735.1 | - | 4 | 357 | |||
DC422585.1 | - | 4 | 373 | |||
DR423861.1 | - | 4 | 296 |