Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCARF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153334.6 | NP_699165.3 | 10 | 1771 | ||
NM_182895.4 | NP_878315.2 | 10 | 1756 | |||
XM_017029065.1 | XP_016884554.1 | 10 | 1802 | |||
GenBank mRNA | AB024433.1 | - | 10 | 1722 | ||
AB052951.1 | - | 10 | 1662 | |||
AF522196.1 | - | 10 | 1771 | |||
BC009326.2 | - | 7 | 839 |