Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004085.3 | NP_004076.1 | 1, 2 | 434 | ||
GenBank mRNA | AK312117.1 | - | 1, 2 | 182 | ||
BC006994.1 | - | 1, 2 | 184 | 4051368 | ||
BC015093.1 | - | 1, 2 | 187 | 3684305 | ||
BC070284.1 | - | 1, 2 | 167 | 4281013 | ||
CN410182.1 | - | 1, 2 | 434 | |||
U66035.1 | - | 1, 2 | 164 |