Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SESTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178123.4 | NP_835224.3 | 12 | 1390 | ||
XM_011512139.2 | XP_011510441.1 | 12 | 1418 | |||
XM_011512141.2 | XP_011510443.1 | 10 | 1095 | |||
XM_017005250.1 | XP_016860739.1 | 12 | 1367 | |||
XM_017005251.1 | XP_016860740.1 | 13 | 1596 | |||
GenBank mRNA | AK096232.1 | - | 12 | 1368 | ||
AL122046.1 | - | 2 | 177 | |||
AY209190.1 | - | 12 | 1287 | |||
BC047578.1 | - | 12 | 1539 | 4815542 |