Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC74B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258307.1 | NP_001245236.1 | 3 | 445 | ||
NM_207310.2 | NP_997193.1 | 3 | 643 | |||
XM_005263840.2 | XP_005263897.1 | 3 | 668 | |||
XM_005263842.2 | XP_005263899.1 | 3 | 470 | |||
XM_006712833.2 | XP_006712896.1 | 3 | 668 | |||
XM_006712834.3 | XP_006712897.1 | 3 | 608 | |||
XM_006712835.2 | XP_006712898.1 | 3 | 470 | |||
XM_006712837.2 | XP_006712900.1 | 3 | 410 | |||
XM_011512142.2 | XP_011510444.1 | 1 | 911 | |||
XM_011512147.1 | XP_011510449.1 | 3 | 668 | |||
GenBank mRNA | AK057609.1 | - | 3 | 567 | ||
AK297246.1 | - | 2 | 1555 | |||
AL133619.1 | - | 1 | 857 | |||
BC067771.1 | - | 3 | 390 | 4824929 |
Gene Symbol: CCDC74A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB299086.1 | - | 3 | 683 | ||
AB299087.1 | - | 3 | 557 | |||
AK097664.1 | - | 1 | 1125 | |||
DA375407.1 | - | 3 | 444 |