Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017919.1 | NP_001017919.1 | 8 | 1255 | ||
NM_033544.2 | NP_291022.2 | 9 | 1339 | |||
GenBank mRNA | AK092661.1 | - | 1 | 770 | ||
AL110105.1 | - | 1 | 271 | |||
BC064606.1 | - | 6 | 1266 | |||
BC094739.1 | - | 6 | 2631 | |||
BC113826.1 | - | 6 | 663 | |||
BC140708.1 | - | 8 | 1253 | 9021576 | ||
BC140709.1 | - | 8 | 1253 | 9021577 |