Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKDCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138370.2 | NP_612379.2 | 3, 4 | 1214 | ||
GenBank mRNA | BC007901.2 | - | 1, 2 | 169 | ||
BC062988.1 | - | 1, 2 | 169 | |||
BC094697.1 | - | 3, 4 | 622 | 30529646 | ||
BC110513.1 | - | 3, 4 | 651 | 40031755 | ||
BC110514.1 | - | 3, 4 | 651 | 40031756 | ||
CF131963.1 | - | 3, 4 | 710 |