Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277126.1 | NP_001264055.1 | 10 | 3347 | ||
NM_001277129.1 | NP_001264058.1 | 9 | 3173 | |||
NM_144687.3 | NP_653288.1 | 10 | 3344 | |||
XM_011527479.1 | XP_011525781.1 | 9 | 3176 | |||
XM_011527480.1 | XP_011525782.1 | 9 | 3176 | |||
XM_011527482.1 | XP_011525784.1 | 8 | 3005 | |||
XM_017027460.1 | XP_016882949.1 | 10 | 3344 | |||
XM_017027461.1 | XP_016882950.1 | 9 | 3176 | |||
XM_017027462.1 | XP_016882951.1 | 9 | 3173 | |||
XM_017027463.1 | XP_016882952.1 | 11 | 3120 | |||
XM_017027464.1 | XP_016882953.1 | 10 | 2918 | |||
XM_017027465.1 | XP_016882954.1 | 11 | 3128 | |||
XM_017027466.1 | XP_016882955.1 | 11 | 3125 | |||
XM_017027467.1 | XP_016882956.1 | 10 | 2920 | |||
GenBank mRNA | AF231021.1 | - | 7 | 2101 | ||
AK095460.1 | - | 9 | 3088 | |||
AK290772.1 | - | 10 | 3126 | |||
AY095146.1 | - | 10 | 3291 | |||
AY116204.1 | - | 10 | 3338 | |||
AY116205.1 | - | 9 | 3170 | |||
AY116206.1 | - | 8 | 3002 | |||
AY116207.1 | - | 7 | 2828 | |||
AY154467.1 | - | 10 | 3291 | |||
BC028069.1 | - | 10 | 3234 | 5212737 |