Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELMSAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001043318.2 | NP_001036783.1 | 10 | 3117 | ||
NM_194278.3 | NP_919254.2 | 10 | 3496 | |||
XM_005268204.1 | XP_005268261.1 | 10 | 3496 | |||
XM_005268205.1 | XP_005268262.1 | 10 | 3052 | |||
XM_005268206.1 | XP_005268263.1 | 11 | 3269 | |||
GenBank mRNA | AK127785.1 | - | 11 | 3495 | ||
BC003121.1 | - | 10 | 2289 | |||
BC006451.1 | - | 10 | 1951 | |||
BC006511.2 | - | 10 | 2747 | |||
BC009202.2 | - | 11 | 3117 | |||
BC015668.2 | - | 10 | 3130 | |||
BC025330.1 | - | 10 | 2747 |