Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153369.2 | NP_699200.2 | 2 | 479 | ||
XM_017011483.1 | XP_016866972.1 | 2 | 479 | |||
GenBank mRNA | AK093136.1 | - | 2 | 438 | ||
AK292735.1 | - | 2 | 408 | |||
AK315016.1 | - | 2 | 401 | |||
BC036115.1 | - | 2 | 388 | 5298999 | ||
DB024559.1 | - | 2 | 438 | |||
DB027926.1 | - | 2 | 481 |