Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317899.1 | NP_001304828.1 | 8 | 1016 | ||
NM_152272.4 | NP_689485.1 | 9 | 1730 | |||
XM_017013961.1 | XP_016869450.1 | 8 | 1186 | |||
XM_017013962.1 | XP_016869451.1 | 10 | 1867 | |||
XM_017013963.1 | XP_016869452.1 | 8 | 993 | |||
XM_017013964.1 | XP_016869453.1 | 9 | 2879 | |||
GenBank mRNA | AK021776.1 | - | 8 | 1159 | ||
AK092500.1 | - | 8 | 966 | |||
AK097202.1 | - | 9 | 1727 | |||
AK315664.1 | - | 8 | 1167 | |||
AL833843.1 | - | 8 | 1135 | |||
BC004344.1 | - | 8 | 958 | |||
BC019110.1 | - | 8 | 1132 | 5093453 | ||
BC042050.1 | - | 9 | 1711 | 5551762 |