Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other WDR20 SIRNA Assays ›
Gene Symbol:
WDR20
Gene Name:
WD repeat domain 20
Gene Aliases:
DMR
Chromosome Location:
Chr.14: 102139518 - 102224686 on Build GRCh38
UniGene ID:
Hs.36859
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s40793
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: WDR20
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242414.1
NP_001229343.1
3
425
NM_001242416.1
NP_001229345.1
2
412
NM_001242417.1
NP_001229346.1
4
688
NM_001242418.1
NP_001229347.1
3
631
NM_144574.3
NP_653175.2
3
595
NM_181291.2
NP_851808.1
3
595
NM_181308.2
NP_851825.1
2
412
XM_006720308.1
XP_006720371.1
2
404
XM_006720309.1
XP_006720372.1
3
378
XM_006720310.1
XP_006720373.1
3
339
XM_006720311.1
XP_006720374.1
2
168
XM_011537335.2
XP_011535637.1
4
707
XM_011537336.2
XP_011535638.1
4
707
XM_011537337.2
XP_011535639.1
4
707
XM_011537338.1
XP_011535640.1
4
654
XM_011537339.2
XP_011535641.1
4
707
XM_011537340.2
XP_011535642.1
4
707
XM_011537341.2
XP_011535643.1
3
613
XM_011537343.1
XP_011535645.1
5
682
XM_011537344.2
XP_011535646.1
4
600
XM_011537345.1
XP_011535647.1
3
378
XM_011537346.1
XP_011535648.1
3
404
XM_011537347.1
XP_011535649.1
4
402
XM_017021766.1
XP_016877255.1
4
654
XM_017021767.1
XP_016877256.1
3
613
XM_017021768.1
XP_016877257.1
3
628
XM_017021770.1
XP_016877259.1
3
628
XM_017021771.1
XP_016877260.1
4
663
XM_017021772.1
XP_016877261.1
4
416
XM_017021773.1
XP_016877262.1
5
509
XM_017021774.1
XP_016877263.1
4
458
XM_017021775.1
XP_016877264.1
4
464
XM_017021776.1
XP_016877265.1
3
378
XM_017021777.1
XP_016877266.1
3
346
XM_017021778.1
XP_016877267.1
4
458
XM_017021779.1
XP_016877268.1
2
168
XM_017021780.1
XP_016877269.1
4
579
XM_017021785.1
XP_016877274.1
4
654
GenBank mRNA
AB209148.1
-
1
4684
AF327354.1
-
3
520
AK056330.1
-
3
337
AK090978.1
-
3
366
AK225852.1
-
2
371
AK297727.1
-
4
626
AL832242.1
-
3
364
AL832500.1
-
3
570
BC028387.2
-
3
554
4823662
BX248274.1
-
2
314
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_027149, BC026761, AK136417, AK052481, AK145389, and others.
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-lxknh:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0