Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: INTS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033547.3 | NP_291025.3 | 17, 18 | 2123 | ||
XM_011545352.2 | XP_011543654.1 | 18, 19 | 2292 | |||
XM_017018560.1 | XP_016874049.1 | 17, 18 | 2156 | |||
GenBank mRNA | AF172822.1 | - | 4, 5 | 509 | ||
AK290866.1 | - | 17, 18 | 2119 | |||
AL834171.1 | - | 17, 18 | 2066 | |||
BC006369.1 | - | 5, 6 | 492 | |||
BC008013.1 | - | 13, 14 | 1576 | |||
BC009859.2 | - | 7, 8 | 782 | |||
BC015664.2 | - | 18, 19 | 2433 | 4554197 |