Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017920.2 | NP_001017920.2 | 3, 4 | 271 | ||
GenBank mRNA | AF086541.1 | - | 3, 4 | 259 | ||
AY324399.1 | - | 3, 4 | 271 | |||
BC127682.1 | - | 3, 4 | 256 | 40127452 | ||
BC127683.1 | - | 3, 4 | 256 | 40127454 | ||
BC140719.1 | - | 3, 4 | 258 | 9021587 | ||
CD675155.1 | - | 3, 4 | 250 | |||
HQ179934.1 | - | 3, 4 | 230 | |||
HQ179939.1 | - | 2, 3 | 169 |