Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017920.2 | NP_001017920.2 | 3 | 237 | ||
GenBank mRNA | AF086541.1 | - | 3 | 225 | ||
AY324399.1 | - | 3 | 237 | |||
BC127683.1 | - | 3 | 222 | 40127454 | ||
CD675155.1 | - | 3 | 216 | |||
HQ179934.1 | - | 3 | 196 | |||
HQ179935.1 | - | 3 | 196 | |||
HQ179936.1 | - | 3 | 196 | |||
HQ179937.1 | - | 3 | 196 | |||
HQ179938.1 | - | 3 | 196 |