Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMBRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007527.1 | NP_001007528.1 | 9 | 1504 | ||
XM_011514162.2 | XP_011512464.1 | 9 | 1946 | |||
XM_017010024.1 | XP_016865513.1 | 10 | 1622 | |||
GenBank mRNA | AK091295.1 | - | 9 | 1444 | ||
AK300433.1 | - | 10 | 1414 | |||
AK307575.1 | - | 9 | 1450 | |||
BC110506.2 | - | 9 | 1443 | 40028737 | ||
BX647340.1 | - | 9 | 1479 | |||
CR749399.1 | - | 9 | 1504 |