Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NQO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000903.2 | NP_000894.1 | 2, 3 | 357 | ||
NM_001025433.1 | NP_001020604.1 | 2, 3 | 357 | |||
NM_001025434.1 | NP_001020605.1 | 2, 3 | 357 | |||
NM_001286137.1 | NP_001273066.1 | 2, 3 | 395 | |||
GenBank mRNA | AK223050.1 | - | 2, 3 | 269 | ||
AK297125.1 | - | 2, 3 | 287 | |||
AK297979.1 | - | 2, 3 | 287 | |||
AK298896.1 | - | 3, 4 | 207 | |||
AK307533.1 | - | 2, 3 | 219 | |||
AK309513.1 | - | 2, 4 | 171 | |||
AK312368.1 | - | 2, 3 | 283 | |||
AK316246.1 | - | 2, 3 | 180 | |||
BC007659.2 | - | 2, 3 | 283 | 3349257 | ||
BC107739.1 | - | 2, 3 | 291 | |||
BE903613.1 | - | 2, 3 | 305 | |||
BM452267.1 | - | 568 | ||||
CD014005.1 | - | 2, 3 | 291 | |||
DC331440.1 | - | 2, 3 | 395 | |||
J03934.1 | - | 2, 3 | 216 |