Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRADA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003786.2 | NP_001003786.1 | 4 | 445 | ||
NM_001003787.2 | NP_001003787.1 | 6 | 556 | |||
NM_001003788.2 | NP_001003788.1 | 5 | 527 | |||
NM_001165969.1 | NP_001159441.1 | 4 | 469 | |||
NM_001165970.1 | NP_001159442.1 | 4 | 424 | |||
NM_153335.5 | NP_699166.2 | 4 | 445 | |||
XM_005257797.2 | XP_005257854.1 | 6 | 407 | |||
XM_005257798.2 | XP_005257855.1 | 4 | 369 | |||
XM_005257799.2 | XP_005257856.1 | 5 | 378 | |||
XM_005257800.2 | XP_005257857.1 | 6 | 431 | |||
XM_005257801.4 | XP_005257858.1 | 6 | 431 | |||
XM_005257803.4 | XP_005257860.1 | 6 | 431 | |||
XM_011525466.2 | XP_011523768.1 | 6 | 431 | |||
XM_011525467.2 | XP_011523769.1 | 6 | 458 | |||
XM_017025312.1 | XP_016880801.1 | 4 | 320 | |||
XM_017025313.1 | XP_016880802.1 | 5 | 402 | |||
XM_017025314.1 | XP_016880803.1 | 4 | 369 | |||
GenBank mRNA | AF308302.1 | - | 6 | 395 | ||
AK074771.1 | - | 4 | 311 | |||
AK075005.1 | - | 4 | 274 | |||
AK293160.1 | - | 4 | 332 | |||
AK301331.1 | - | 4 | 296 | |||
AL832407.1 | - | 5 | 340 | |||
AY290821.2 | - | 4 | 445 | |||
BC043641.1 | - | 5 | 318 | 3908187 | ||
BG339051.1 | - | 331 |