Gene Symbol: TIMM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001563.2 | NP_001001563.1 | 3 | 713 | ||
XM_011527491.2 | XP_011525793.1 | 3 | 256 | |||
XM_017027472.1 | XP_016882961.1 | 3 | 279 | |||
GenBank mRNA | AY444561.1 | - | 3 | 247 | ||
AY551341.1 | - | 3 | 271 | |||
BC009072.1 | - | 3 | 245 | |||
BC010736.1 | - | 3 | 266 | |||
BC050082.1 | - | 3 | 289 | |||
BC121146.1 | - | 3 | 641 | 40121256 | ||
BC121147.1 | - | 3 | 641 | 40121258 | ||
BQ925812.1 | - | 3 | 300 |