Search Thermo Fisher Scientific
Gene Symbol: TIFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052864.2 | NP_443096.1 | 2 | 449 | ||
GenBank mRNA | AB062110.1 | - | 1 | 167 | ||
AB097011.1 | - | 2 | 451 | |||
AB097038.1 | - | 2 | 451 | |||
AK292900.1 | - | 2 | 446 | |||
BC008294.1 | - | 2 | 433 | |||
BC014259.2 | - | 2 | 429 | 4763971 |