Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139072.3 | NP_620711.3 | 5 | 1095 | ||
XM_005246950.2 | XP_005247007.1 | 5 | 1082 | |||
GenBank mRNA | AF442487.1 | - | 5 | 1008 | ||
AK289553.1 | - | 5 | 1053 | |||
AK304596.1 | - | 3 | 445 | |||
AK304631.1 | - | 3 | 474 | |||
AY358891.1 | - | 5 | 1058 | |||
BC024766.2 | - | 1 | 101 | |||
BC035009.1 | - | 5 | 1085 | 4820343 |