Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAPRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286829.1 | NP_001273758.1 | 13 | 1636 | ||
NM_145201.5 | NP_660202.3 | 13 | 1675 | |||
XM_017013975.1 | XP_016869464.1 | 11 | 3322 | |||
XM_017013976.1 | XP_016869465.1 | 12 | 3231 | |||
XM_017013977.1 | XP_016869466.1 | 11 | 3022 | |||
XM_017013978.1 | XP_016869467.1 | 10 | 2974 | |||
XM_017013979.1 | XP_016869468.1 | 11 | 1518 | |||
XM_017013980.1 | XP_016869469.1 | 12 | 1538 | |||
GenBank mRNA | AB242230.1 | - | 13 | 1662 | ||
AF258565.1 | - | 7 | 1123 | |||
AK097002.1 | - | 13 | 1667 | |||
AY214325.1 | - | 12 | 1750 | |||
AY214326.1 | - | 13 | 1663 | |||
AY214327.1 | - | 13 | 1624 | |||
BC006284.2 | - | 13 | 1647 | 3957135 | ||
BC014190.2 | - | 11 | 1829 | |||
BC032466.1 | - | 8 | 961 | 5210589 |