Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNA2D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172364.4 | NP_758952.4 | 5 | 869 | ||
XM_011521041.2 | XP_011519343.1 | 5 | 4771 | |||
GenBank mRNA | AF516695.1 | - | 5 | 1001 | ||
BC150186.1 | - | 5 | 802 | 40148321 | ||
BX537436.1 | - | 5 | 832 | |||
BX537437.1 | - | 5 | 870 |