Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNA2D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172364.4 | NP_758952.4 | 29 | 2908 | ||
XM_011521041.2 | XP_011519343.1 | 28 | 6747 | |||
GenBank mRNA | AF516695.1 | - | 29 | 3040 | ||
AK301234.1 | - | 2 | 149 | |||
AL137658.1 | - | 7 | 518 | |||
BC048288.1 | - | 2 | 85 | |||
BC150186.1 | - | 28 | 2831 | 40148321 | ||
BI908421.1 | - | 2 | 115 | |||
BX537436.1 | - | 29 | 2871 | |||
BX537437.1 | - | 29 | 2909 |