Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033256.2 | NP_150281.1 | 2 | 473 | ||
GenBank mRNA | AB056508.1 | - | 2 | 414 | ||
AF123763.1 | - | 2 | 264 | |||
AY050670.1 | - | 2 | 258 | |||
BC021089.1 | - | 2 | 238 | 5019808 |