Search Thermo Fisher Scientific
Gene Symbol: KIF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138424.1 | NP_612433.1 | 10 | 1095 | ||
XM_005251683.4 | XP_005251740.1 | 13 | 2084 | |||
XM_006716947.2 | XP_006717010.1 | 12 | 1336 | |||
XM_011518190.2 | XP_011516492.1 | 13 | 2119 | |||
XM_011518191.2 | XP_011516493.1 | 13 | 2118 | |||
XM_011518192.2 | XP_011516494.1 | 13 | 2116 | |||
XM_011518193.2 | XP_011516495.1 | 12 | 1969 | |||
XM_017014240.1 | XP_016869729.1 | 7 | 775 | |||
GenBank mRNA | BC010626.1 | - | 10 | 1095 | 3865868 |