Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LARP4 SIRNA Assays ›
Gene Symbol:
LARP4
Gene Name:
La ribonucleoprotein domain family member 4
Gene Aliases:
PP13296
Chromosome Location:
Chr.12: 50400809 - 50480005 on Build GRCh38
UniGene ID:
Hs.26613
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s41455
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LARP4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170803.1
NP_001164274.1
6
752
NM_001170804.1
NP_001164275.1
6
752
NM_001170808.1
NP_001164279.1
6
752
NM_052879.4
NP_443111.4
6
752
NM_199188.2
NP_954658.2
6
749
NM_199190.2
NP_954660.1
6
752
NR_033200.1
7
801
XM_005268613.2
XP_005268670.1
7
801
XM_011537828.2
XP_011536130.1
7
856
XM_011537829.2
XP_011536131.1
7
837
XM_011537830.2
XP_011536132.1
6
684
XM_011537831.2
XP_011536133.1
6
748
XM_011537833.2
XP_011536135.1
7
856
XM_011537834.2
XP_011536136.1
7
857
XM_011537835.2
XP_011536137.1
7
837
XM_011537836.2
XP_011536138.1
7
838
XM_011537838.2
XP_011536140.1
6
596
XM_011537839.2
XP_011536141.1
7
740
XM_011537840.2
XP_011536142.1
7
793
XM_011537841.2
XP_011536143.1
6
646
XM_011537843.2
XP_011536145.1
7
838
XM_011537844.2
XP_011536146.1
7
857
XM_017018750.1
XP_016874239.1
6
678
XM_017018751.1
XP_016874240.1
7
617
XM_017018752.1
XP_016874241.1
6
600
XM_017018753.1
XP_016874242.1
6
633
XM_017018754.1
XP_016874243.1
7
783
XM_017018755.1
XP_016874244.1
6
663
XM_017018756.1
XP_016874245.1
7
715
XM_017018757.1
XP_016874246.1
8
885
XM_017018758.1
XP_016874247.1
7
821
XM_017018759.1
XP_016874248.1
6
646
XM_017018760.1
XP_016874249.1
6
752
XM_017018761.1
XP_016874250.1
6
745
XM_017018762.1
XP_016874251.1
6
745
GenBank mRNA
AF370416.1
-
6
630
AK125113.1
-
5
542
AK291746.1
-
6
621
AL832198.1
-
6
695
AL833272.1
-
6
662
AY004310.1
-
7
656
BC022377.1
-
6
654
BC063479.1
-
6
653
5219803
BX647457.1
-
6
576
CR936626.1
-
6
670
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
regulation of cell morphogenesis
nucleotide binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0