Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC52A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033409.3 | NP_212134.3 | 2 | 763 | ||
XM_005260655.3 | XP_005260712.1 | 3 | 692 | |||
XM_011529148.1 | XP_011527450.1 | 3 | 1201 | |||
GenBank mRNA | AK074650.1 | - | 2 | 663 | ||
AK291706.1 | - | 2 | 681 | |||
BC009750.2 | - | 2 | 666 | 3689286 | ||
JX478249.1 | - | 1 | 424 |
Set Membership: |
![]() |