Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052920.1 | NP_443152.1 | 13 | 2894 | ||
XM_006711929.3 | XP_006711992.1 | 12 | 3990 | |||
XM_011532501.2 | XP_011530803.1 | 10 | 1573 | |||
XM_017003264.1 | XP_016858753.1 | 12 | 2794 | |||
GenBank mRNA | AB067508.1 | - | 7 | 4197 | ||
BC013982.1 | - | 10 | 1883 |