Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other OSBPL9 SIRNA Assays ›
Gene Symbol:
OSBPL9
Gene Name:
oxysterol binding protein like 9
Gene Aliases:
ORP-9, ORP9
Chromosome Location:
Chr.1: 51616874 - 51789219 on Build GRCh38
UniGene ID:
Hs.21938
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s41695
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: OSBPL9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_024586.5
NP_078862.4
14, 15
1399
NM_148904.3
NP_683702.1
10, 11
1116
NM_148905.3
NP_683703.1
9, 10
1020
NM_148906.2
NP_683704.2
13, 14
1348
NM_148907.2
NP_683705.1
10, 11
993
NM_148908.3
NP_683706.3
13, 14
1360
NM_148909.3
NP_683707.3
14, 15
1429
NR_036662.1
13, 14
1320
XM_006710318.2
XP_006710381.1
13, 14
1159
XM_006710320.3
XP_006710383.1
11, 12
996
XM_006710321.2
XP_006710384.1
10, 11
992
XM_006710322.3
XP_006710385.1
11, 12
1153
XM_006710323.2
XP_006710386.1
10, 11
1061
XM_006710324.3
XP_006710387.1
12, 13
1071
XM_006710325.3
XP_006710388.1
11, 12
1041
XM_006710326.2
XP_006710389.1
9, 10
1043
XM_011540599.2
XP_011538901.1
15, 16
1253
XM_011540600.2
XP_011538902.1
14, 15
1220
XM_011540601.2
XP_011538903.1
13, 14
1181
XM_011540602.2
XP_011538904.1
14, 15
1155
XM_011540603.2
XP_011538905.1
14, 15
1179
XM_011540604.2
XP_011538906.1
11, 12
954
XM_017000218.1
XP_016855707.1
12, 13
1120
XM_017000219.1
XP_016855708.1
13, 14
1140
XM_017000220.1
XP_016855709.1
10, 11
881
XM_017000221.1
XP_016855710.1
10, 11
832
XM_017000222.1
XP_016855711.1
14, 15
1365
XM_017000223.1
XP_016855712.1
14, 15
1363
XM_017000224.1
XP_016855713.1
11, 12
1308
XM_017000225.1
XP_016855714.1
12, 13
1159
GenBank mRNA
AF392445.1
-
14, 15
1173
AK022554.1
-
9, 10
800
AK027535.1
-
13, 14
1130
AK027707.1
-
10, 11
956
AK056617.1
-
14, 15
1218
AK091703.1
-
13, 14
1102
AK124037.1
-
5, 6
1646
AK128043.1
-
16, 17
1623
AK297656.1
-
11, 12
1146
AK315167.1
-
10, 11
992
AY178997.1
-
13, 14
1122
BC025978.2
-
10, 11
1026
3621193
CR457288.1
-
7, 8
667
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00541027_m1
Gene Ontology Categories:
Function(s)
Process(es)
lipid transport
lipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbcvc:80/10.239.164.135:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline