Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022776.4 | NP_073613.2 | 7 | 1739 | ||
GenBank mRNA | AF392454.1 | - | 7 | 1310 | ||
AK023074.1 | - | 1 | 136 | |||
AK023226.1 | - | 2 | 169 | |||
AK292702.1 | - | 7 | 1091 | |||
BC010422.1 | - | 2 | 323 | |||
BC065213.1 | - | 7 | 1089 | 3916115 |