Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022776.4 | NP_073613.2 | 6 | 1440 | ||
GenBank mRNA | AF346292.1 | - | 6 | 867 | ||
AF392454.1 | - | 6 | 1011 | |||
AK292702.1 | - | 6 | 792 | |||
BC010422.1 | - | 1 | 24 | |||
BC065213.1 | - | 6 | 790 | 3916115 |