Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052964.2 | NP_443196.2 | 6 | 412 | ||
XM_011513775.2 | XP_011512077.1 | 6 | 643 | |||
XM_017007684.1 | XP_016863173.1 | 6 | 643 | |||
XM_017007685.1 | XP_016863174.1 | 6 | 643 | |||
GenBank mRNA | AB032369.1 | - | 4 | 221 | ||
AB110420.1 | - | 6 | 515 | |||
AK093001.1 | - | 6 | 414 | |||
BC029887.1 | - | 6 | 442 | |||
BC137553.1 | - | 6 | 341 | 9021553 | ||
BC137556.1 | - | 6 | 341 | 9021556 |