Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRGPRD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198923.2 | NP_944605.2 | 1 | 47 | ||
GenBank mRNA | AB154410.1 | - | 1 | 47 | ||
AY427820.1 | - | 1 | 47 |
Set Membership: |
![]() |