Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300922.1 | NP_001287851.1 | 2 | 232 | ||
NM_054113.3 | NP_473454.1 | 4 | 379 | |||
GenBank mRNA | AB050868.1 | - | 4 | 329 | ||
BC069428.1 | - | 2 | 182 | 7262131 | ||
BC069524.1 | - | 4 | 310 | |||
BC112200.1 | - | 4 | 379 | 8327668 | ||
BC113591.1 | - | 4 | 379 | 8322643 |