Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288821.1 | NP_001275750.1 | 6 | 1013 | ||
NM_001288823.1 | NP_001275752.1 | 7 | 1325 | |||
NM_058170.3 | NP_477518.2 | 6 | 1264 | |||
NR_110210.1 | 8 | 2062 | ||||
NR_110211.1 | 7 | 2001 | ||||
NR_110212.1 | 7 | 1750 | ||||
XM_017000240.1 | XP_016855729.1 | 7 | 1168 | |||
GenBank mRNA | AF397392.2 | - | 6 | 906 | ||
AF397393.2 | - | 7 | 1643 | |||
AF397394.1 | - | 6 | 935 | |||
AF397395.1 | - | 7 | 1672 | |||
AF397396.1 | - | 7 | 996 | |||
AF397397.1 | - | 8 | 1733 | |||
AK095551.1 | - | 4 | 490 | |||
AK095724.1 | - | 6 | 1060 | |||
AK300870.1 | - | 6 | 1013 | |||
AY358722.1 | - | 6 | 977 | |||
BC022531.1 | - | 6 | 1022 | 4812035 |