Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZFYVE27 SIRNA Assays ›
Gene Symbol:
ZFYVE27
Gene Name:
zinc finger FYVE-type containing 27
Gene Aliases:
PROTRUDIN, SPG33
Chromosome Location:
Chr.10: 97737121 - 97760907 on Build GRCh38
UniGene ID:
Hs.744075
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s42248
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZFYVE27
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002261.3
NP_001002261.1
1
179
NM_001002262.3
NP_001002262.1
2
384
NM_001174120.1
NP_001167591.1
2
380
NM_001174122.1
NP_001167593.1
2
380
NM_144588.6
NP_653189.3
2
384
XM_005269502.3
XP_005269559.1
2
373
XM_005269503.3
XP_005269560.1
2
377
XM_005269504.3
XP_005269561.1
2
373
XM_005269505.4
XP_005269562.1
2
373
XM_005269506.3
XP_005269563.1
2
373
XM_005269508.3
XP_005269565.1
2
373
XM_005269509.3
XP_005269566.1
2
377
XM_005269510.3
XP_005269567.1
2
376
XM_005269511.3
XP_005269568.1
2
382
XM_011539252.2
XP_011537554.1
2
373
XM_011539253.2
XP_011537555.1
2
373
XM_017015644.1
XP_016871133.1
2
373
XM_017015645.1
XP_016871134.1
2
373
XM_017015646.1
XP_016871135.1
2
373
XM_017015647.1
XP_016871136.1
2
377
XM_017015648.1
XP_016871137.1
2
376
XM_017015649.1
XP_016871138.1
2
376
XM_017015651.1
XP_016871140.1
2
373
XM_017015652.1
XP_016871141.1
2
377
XM_017015653.1
XP_016871142.1
2
373
XM_017015654.1
XP_016871143.1
2
377
XM_017015655.1
XP_016871144.1
2
382
GenBank mRNA
AK074876.1
-
2
365
AK097945.1
-
2
373
AK299735.1
-
2
373
AK300438.1
-
2
377
AL561314.3
-
2
296
AL834235.1
-
1
113
BC030621.2
-
2
377
4818199
BF508037.1
-
2
347
BI758753.1
-
2
372
BX647836.1
-
2
319
CB150532.1
-
2
373
DC302372.1
-
2
380
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
neuron projection development
neurotrophin TRK receptor signaling pathway
protein localization to plasma membrane
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0