Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004947.4 | NP_004938.1 | 12 | 999 | ||
XM_005264914.3 | XP_005264971.1 | 12 | 1271 | |||
XM_005264915.3 | XP_005264972.1 | 12 | 1271 | |||
XM_005264916.3 | XP_005264973.1 | 12 | 1049 | |||
XM_005264917.3 | XP_005264974.1 | 12 | 1271 | |||
XM_005264918.3 | XP_005264975.1 | 12 | 1048 | |||
XM_006713008.3 | XP_006713071.1 | 12 | 1271 | |||
XM_006713009.3 | XP_006713072.1 | 12 | 1271 | |||
XM_006713010.3 | XP_006713073.1 | 12 | 1034 | |||
XM_017005825.1 | XP_016861314.1 | 12 | 1050 | |||
XM_017005826.1 | XP_016861315.1 | 12 | 1040 | |||
XM_017005827.1 | XP_016861316.1 | 3 | 191 | |||
GenBank mRNA | AB002297.1 | - | 7 | 607 | ||
AY145303.1 | - | 12 | 976 | |||
AY254099.1 | - | 12 | 999 |