Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC36A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286139.1 | NP_001273068.1 | 10 | 1372 | ||
NM_152313.3 | NP_689526.2 | 10 | 1335 | |||
XM_017017175.1 | XP_016872664.1 | 9 | 1252 | |||
XM_017017176.1 | XP_016872665.1 | 9 | 1204 | |||
GenBank mRNA | AK096251.1 | - | 10 | 1371 | ||
AY162216.1 | - | 10 | 1289 | |||
BC047374.1 | - | 10 | 1302 | 5313230 |