Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: OR56A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001917.2 | NP_001001917.2 | 1 | 461 | ||
GenBank mRNA | BC136904.1 | - | 1 | 461 | 9020895 | |
BC136911.1 | - | 1 | 461 | 9020902 |
Set Membership: |
G-protein coupled receptor |