Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018558.3 | NP_061028.3 | 5, 6 | 634 | ||
XM_011531184.2 | XP_011529486.1 | 5, 6 | 922 | |||
GenBank mRNA | AF144648.1 | - | 5, 6 | 599 | ||
AF189259.1 | - | 5, 6 | 634 | |||
BC109210.1 | - | 5, 6 | 634 | 40006890 | ||
BC109211.1 | - | 5, 6 | 634 | 40006899 |