Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139015.4 | NP_620584.2 | 9 | 1343 | ||
XM_011537925.2 | XP_011536227.1 | 9 | 873 | |||
GenBank mRNA | AB252457.1 | - | 9 | 935 | ||
AJ345030.1 | - | 9 | 911 | |||
AJ420898.1 | - | 9 | 882 | |||
AK074916.1 | - | 8 | 826 | |||
AK223590.1 | - | 9 | 1339 | |||
AK291161.1 | - | 9 | 1165 | |||
AY169313.1 | - | 9 | 1117 | |||
AY358767.1 | - | 2 | 189 | |||
BC009551.1 | - | 2 | 211 | |||
BC025781.1 | - | 6 | 618 | |||
BC073910.1 | - | 9 | 1124 | 4538522 | ||
BC101625.1 | - | 9 | 938 | 8069131 | ||
BC101627.1 | - | 9 | 938 | 8069133 |