Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASE11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145250.3 | NP_660293.1 | 3 | 284 | ||
GenBank mRNA | AK098687.1 | - | 2 | 141 | ||
AK292348.1 | - | 2 | 285 | |||
AK314065.1 | - | 3 | 436 | |||
AY358794.1 | - | 3 | 564 | |||
AY665806.1 | - | 1 | 100 | |||
BC025410.1 | - | 3 | 211 | 4826406 | ||
BX161458.1 | - | 3 | 256 | |||
CB995247.1 | - | 3 | 310 | |||
FJ237361.1 | - | 3 | 211 |