Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC25A29 SIRNA Assays ›
Gene Symbol:
SLC25A29
Gene Name:
solute carrier family 25 member 29
Gene Aliases:
C14orf69, CACL, ORNT3
Chromosome Location:
Chr.14: 100277986 - 100306547 on Build GRCh38
UniGene ID:
Hs.578109
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s42614
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC25A29
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039355.2
NP_001034444.1
4
1270
NM_001291813.1
NP_001278742.1
5
1638
NM_001291814.1
NP_001278743.1
5
1342
NM_152333.3
NP_689546.1
3
1226
XM_006720038.1
XP_006720101.1
6
1890
XM_006720039.3
XP_006720102.1
5
1373
XM_011536443.1
XP_011534745.1
7
1946
XM_011536444.1
XP_011534746.1
5
1731
XM_011536446.1
XP_011534748.1
5
2044
XM_011536447.1
XP_011534749.1
4
1982
XM_011536448.1
XP_011534750.1
4
1972
XM_017020979.1
XP_016876468.1
5
1416
XM_017020980.1
XP_016876469.1
5
6544
XM_017020981.1
XP_016876470.1
4
6068
XM_017020982.1
XP_016876471.1
4
4598
XM_017020983.1
XP_016876472.1
4
4588
XM_017020984.1
XP_016876473.1
4
4302
XM_017020985.1
XP_016876474.1
5
4347
XM_017020986.1
XP_016876475.1
4
3783
XM_017020987.1
XP_016876476.1
5
3763
GenBank mRNA
AK095532.1
-
2
1710
AK096294.1
-
3
1118
AK123821.1
-
4
1162
AK172788.1
-
4
1192
AK289627.1
-
3
986
AL832299.1
-
2
3765
BC132964.1
-
2
868
40146986
BC132966.1
-
2
868
40146988
BX247983.1
-
4
1246
Back To Top
More Information
Panther Classification:
Molecular Function -
transporter
Gene Ontology Categories:
Function(s)
Process(es)
translation
acyl carnitine transmembrane transport
structural constituent of ribosome
acyl carnitine transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbzcs:80/10.239.166.68:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline