Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308089.1 | NP_001295018.1 | 4 | 475 | ||
NM_001324130.1 | NP_001311059.1 | 4 | 683 | |||
NM_133450.3 | NP_597707.1 | 5 | 677 | |||
NR_040252.1 | 4 | 680 | ||||
XM_005255119.3 | XP_005255176.1 | 3 | 377 | |||
XM_011522372.1 | XP_011520674.1 | 4 | 607 | |||
XM_011522373.1 | XP_011520675.1 | 4 | 489 | |||
XM_011522378.1 | XP_011520680.1 | 3 | 234 | |||
GenBank mRNA | AB075857.1 | - | 2 | 1113 | ||
AK057329.1 | - | 2 | 278 | |||
AK123496.1 | - | 5 | 678 | |||
AK301683.1 | - | 3 | 305 | |||
BC050387.1 | - | 4 | 316 | 5751728 | ||
BM545219.1 | - | 4 | 526 | |||
DR004376.1 | - | 3 | 413 |