Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324130.1 | NP_001311059.1 | 2, 3 | 404 | ||
NM_133450.3 | NP_597707.1 | 3, 4 | 472 | |||
NR_040252.1 | 2, 3 | 401 | ||||
XM_011522372.1 | XP_011520674.1 | 2, 3 | 402 | |||
XM_011522373.1 | XP_011520675.1 | 2, 3 | 284 | |||
XM_017022936.1 | XP_016878425.1 | 2, 3 | 404 | |||
GenBank mRNA | AK123496.1 | - | 3, 4 | 473 | ||
BM545219.1 | - | 2, 3 | 247 |