Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001154458.2 | NP_001147930.1 | 9 | 1067 | ||
NM_144605.4 | NP_653206.2 | 10 | 1205 | |||
XM_006720846.2 | XP_006720909.1 | 10 | 1341 | |||
XM_011522379.2 | XP_011520681.1 | 9 | 854 | |||
XM_017022938.1 | XP_016878427.1 | 9 | 1319 | |||
GenBank mRNA | AK058139.1 | - | 10 | 1031 | ||
AK098718.1 | - | 10 | 1177 | |||
BC024017.1 | - | 10 | 1003 | |||
BC035619.1 | - | 10 | 1011 | 5172555 | ||
DQ456996.1 | - | 9 | 893 | |||
DQ517531.1 | - | 10 | 1082 | |||
EF620906.1 | - | 10 | 1205 |