Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERICH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002912.4 | NP_001002912.4 | 14 | 4440 | ||
XM_017000274.1 | XP_016855763.1 | 14 | 4440 | |||
XM_017000275.1 | XP_016855764.1 | 14 | 4230 | |||
XM_017000276.1 | XP_016855765.1 | 4 | 2209 | |||
XM_017000277.1 | XP_016855766.1 | 3 | 2167 | |||
XM_017000278.1 | XP_016855767.1 | 4 | 2182 | |||
XM_017000279.1 | XP_016855768.1 | 4 | 2636 | |||
XM_017000280.1 | XP_016855769.1 | 3 | 2577 | |||
XM_017000281.1 | XP_016855770.1 | 2 | 2933 | |||
XM_017000282.1 | XP_016855771.1 | 3 | 2140 | |||
XM_017000283.1 | XP_016855772.1 | 3 | 2594 | |||
XM_017000284.1 | XP_016855773.1 | 2 | 2388 | |||
XM_017000285.1 | XP_016855774.1 | 2 | 2535 | |||
GenBank mRNA | AL834414.1 | - | 1 | 1507 | ||
CR749349.1 | - | 2 | 2363 |