Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147192.2 | NP_671725.1 | 2 | 310 | ||
NM_172225.1 | NP_757379.1 | 2 | 295 | |||
XM_011540668.2 | XP_011538970.1 | 3 | 541 | |||
XM_017000289.1 | XP_016855778.1 | 3 | 333 | |||
GenBank mRNA | AB037699.1 | - | 2 | 295 | ||
AF398527.1 | - | 2 | 286 | |||
AF398528.1 | - | 2 | 301 | |||
AY726596.1 | - | 1 | 120 |