Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001942.3 | NP_001933.2 | 10 | 1605 | ||
GenBank mRNA | AF097935.1 | - | 10 | 1606 | ||
X56654.1 | - | 10 | 1470 |